Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR4D5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599920 Supplier Invitrogen™ Supplier No. PA599920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR4D5.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4D5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR4D5
Gene Accession No. Q8NGN0
Gene Alias Olfactory receptor 4D5; olfactory receptor family 4 subfamily D member 5; olfactory receptor OR11-276; olfactory receptor, family 4, subfamily D, member 5; OR11-276; OR4D5
Gene Symbols OR4D5
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR4D5(Accession Q8NGN0), corresponding to amino acid residues T279-H318.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219875
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.