Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR4D6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309783100UL
Description
OR4D6 Polyclonal specifically detects OR4D6 in Human samples. It is validated for Western Blot.Specifications
OR4D6 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
olfactory receptor 4D6, Olfactory receptor OR11-250, olfactory receptor, family 4, subfamily D, member 6, OR11-250 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR4D6 (NP_001004708). Peptide sequence YCRPFMTLPMDTTISINNTVITPMLNPIIYSLRNQEMKSAMQRLQRRLGP | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
219983 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction