Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR4F15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571521
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571521 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571521 Supplier Thermo Scientific Supplier No. PA571521
Add to Cart
Add to Cart

OR4F15 Polyclonal Antibody

OR4F15 Polyclonal Antibody for Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4F15
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR4F15
Gene Accession No. Q8NGB8
Gene Alias Olfactory receptor 4F15; olfactory receptor family 4 subfamily F member 15; olfactory receptor OR15-14; olfactory receptor, family 4, subfamily F, member 15; OR4F15
Gene Symbols OR4F15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 69-96 amino acids from the N-terminal region of human OR4F15.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.