Learn More
Invitrogen™ OR4K5 Polyclonal Antibody

Description
OR4K5 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | OR4K5 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OR4K5 |
| Gene Accession No. | Q8NGD3 |
| Gene Alias | Olfactory receptor 4K5; olfactory receptor family 4 subfamily K member 5; olfactory receptor OR14-16; olfactory receptor, family 4, subfamily K, member 5; OR14-16; OR4K5 |
| Gene Symbols | OR4K5 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.