Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR4M2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571043
Description
OR4M2 Polyclonal Antibody for Western Blot
OR4M2 is an odorant receptor (Potential).
Specifications
| OR4M2 | |
| Polyclonal | |
| Unconjugated | |
| OR4M2 | |
| Olfactory receptor 4M2; olfactory receptor family 4 subfamily M member 2; olfactory receptor OR15-3; olfactory receptor, family 4, subfamily M, member 2; OR15-3; OR4M2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 390538 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NGB6 | |
| OR4M2 | |
| synthetic peptide directed towards the C-terminal region of human OR4M2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction