Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR4N4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563444 Supplier Invitrogen™ Supplier No. PA563444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEVKTSMKRL LSRHVVCQVD FIIRN Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 52%.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4N4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR4N4
Gene Accession No. Q8N0Y3
Gene Alias Olfactory receptor 4N4; olfactory receptor family 4 subfamily N member 4; olfactory receptor OR15-1; Olfactory receptor OR15-5; olfactory receptor, family 4, subfamily N, member 4; OR15-1; OR15-5; OR4N4
Gene Symbols OR4N4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR4N4. Recombinant protein control fragment (Product #RP-101296).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283694
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.