Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR4S1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549240

Catalog No. PA549240


Only null left
Add to Cart

Description

Description

OR4S1 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OR4S1
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q8NGB4
OR4S1
KLH conjugated synthetic peptide between 282-309 amino acids from the C-terminal region of human OR4S1.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
OR4S1
olfactory receptor 4S1; olfactory receptor family 4 subfamily S member 1; olfactory receptor OR11-100; olfactory receptor, family 4, subfamily S, member 1; OR11-100; OR4S1
Rabbit
Antigen affinity chromatography, Protein A
RUO
256148
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.