Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR51A1P Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100935
Description
Antibody detects endogenous levels of total OR51A1P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
OR51A1P | |
Polyclonal | |
Unconjugated | |
OR51A1P | |
Olfactory receptor 51L1; olfactory receptor 52E2; olfactory receptor family 51 subfamily L member 1; olfactory receptor family 52 subfamily E member 2; olfactory receptor OR11-31; olfactory receptor, family 10, subfamily AC, member 1 pseudogene; olfactory receptor, family 51, subfamily L, member 1; olfactory receptor, family 52, subfamily E, member 2; OR11-31; OR51L1; OR52E2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26537 | |
-20°C | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
0 | |
OR51A1P | |
A synthesized peptide derived from human(unreviewed) OR51A1P(Accession Q9H2C6), corresponding to amino acid residues P16-Y36. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction