Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR51E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535298
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535298 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535298 Supplier Invitrogen™ Supplier No. PA535298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR51E1 Polyclonal Antibody for Western Blot

Odorant receptor (Potential).
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR51E1
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR51E1
Gene Accession No. Q8TCB6
Gene Alias DGPCR; D-GPCR; Dresden-G-protein-coupled receptor; GPR136; GPR164; G-protein coupled receptor 164; Olfactory receptor 51E1; olfactory receptor 52A3; olfactory receptor family 51 subfamily E member 1; olfactory receptor OR11-15; olfactory receptor, family 51, subfamily E, member 1 pseudogene; olfactory receptor, family 52, subfamily A, member 3 pseudogene; OR51E1; OR51E1P; OR52A3P; POGR; prostate overexpressed G protein-coupled receptor; prostate-overexpressed G protein-coupled receptor; Prostate-specific G protein-coupled receptor 2; PSGR2
Gene Symbols OR51E1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 280-309 amino acids from the C-terminal region of human OR51E1.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143503
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.