Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR51M1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100966 Supplier Invitrogen™ Supplier No. PA5100966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR51M1.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR51M1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR51M1
Gene Accession No. Q9H341
Gene Alias HOR5'Beta7; Odorant receptor HOR5'beta7; olfactory receptor 51M1; olfactory receptor family 51 subfamily M member 1; olfactory receptor OR11-40; olfactory receptor, family 51, subfamily M, member 1; OR11-40; OR51M1
Gene Symbols OR51M1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR51M1(Accession Q9H341), corresponding to amino acid residues L257-K307.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390059
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.