Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR51V1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309582100UL
Description
OR51V1 Polyclonal specifically detects OR51V1 in Human samples. It is validated for Western Blot.Specifications
| OR51V1 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| Odorant receptor HOR3'beta1, Olfactory receptor 51A12, olfactory receptor 51V1, Olfactory receptor OR11-36, olfactory receptor, family 51, subfamily V, member 1, OR11-36, OR51A12 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR51V1 (NP_001004760). Peptide sequence YYALMLVICILLLDAILILFSYILILKSVLAVASQEERHKLFQTCISHIC | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 283111 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction