Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR52A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309552100UL
Description
OR52A1 Polyclonal specifically detects OR52A1 in Human samples. It is validated for Western Blot.Specifications
OR52A1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
HPFH1OR, odorant receptor HOR3'beta4, olfactory receptor 52A1, olfactory receptor OR11-319, olfactory receptor, family 52, subfamily A, member 1 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR52A1 (NP_036507). Peptide sequence IQIFITVFRLPQKEARFKAFNTCIAHICVFLQFYLLAFFSFFTHRFGSHI | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
23538 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction