Learn More
OR52I1 Polyclonal Antibody

Description
OR52I1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | OR52I1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OR52I1 |
| Gene Accession No. | Q8NGK6 |
| Gene Alias | Olfactory receptor 52I1; olfactory receptor family 52 subfamily I member 1; olfactory receptor OR11-13; olfactory receptor, family 52, subfamily I, member 1; OR11-13; OR52I1 |
| Gene Symbols | OR52I1 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.