Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR52M1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599942
Description
Antibody detects endogenous levels of total OR52M1.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
OR52M1 | |
Polyclonal | |
Unconjugated | |
OR52M1 | |
Olfactory receptor 52M1; olfactory receptor family 52 subfamily M member 1; olfactory receptor OR11-11; olfactory receptor, family 52, subfamily M, member 1 pseudogene; olfactory receptor, family 52, subfamily M, member 3 pseudogene; OR11-11; OR52M1; OR52M1P; OR52M3P | |
Rabbit | |
Affinity chromatography | |
RUO | |
119772 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NGK5 | |
OR52M1 | |
A synthesized peptide derived from human OR52M1(Accession Q8NGK5), corresponding to amino acid residues L210-V260. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction