Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR52N1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24911025UL
Description
OR52N1 Polyclonal antibody specifically detects OR52N1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
OR52N1 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
olfactory receptor 52N1, Olfactory receptor OR11-61, olfactory receptor, family 52, subfamily N, member 1, OR11-61 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TRQVRESVIRFFLKGKDNSHNF | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79473 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction