Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR52N1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2833270.1ML
Description
OR52N1 Polyclonal specifically detects OR52N1 in Human samples. It is validated for Western Blot.Specifications
OR52N1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79473 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
olfactory receptor 52N1, Olfactory receptor OR11-61, olfactory receptor, family 52, subfamily N, member 1, OR11-61 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR52N1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction