Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR52N5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599944
Description
Antibody detects endogenous levels of total OR52N5.
Odorant receptor.
Specifications
OR52N5 | |
Polyclonal | |
Unconjugated | |
OR52N5 | |
Olfactory receptor 52N5; olfactory receptor family 52 subfamily N member 5; olfactory receptor OR11-62; olfactory receptor, family 52, subfamily N, member 5; OR11-62; OR52N5; OR52N5Q | |
Rabbit | |
Affinity chromatography | |
RUO | |
390075 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NH56 | |
OR52N5 | |
A synthesized peptide derived from human OR52N5(Accession Q8NH56), corresponding to amino acid residues Y290-G324. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction