Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR52Z1P Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5100936
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100936 Supplier Thermo Scientific Supplier No. PA5100936

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR52Z1P.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR52Z1P
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR52Z1
Gene Accession No. P0C646
Gene Alias Odorant receptor HOR3'beta5; olfactory receptor 52A5; Olfactory receptor 52B2; olfactory receptor 52K1; Olfactory receptor 52N4; olfactory receptor 52N5; olfactory receptor family 52 subfamily A member 5; olfactory receptor family 52 subfamily B member 2; olfactory receptor family 52 subfamily K member 1; olfactory receptor family 52 subfamily N member 4 (gene/pseudogene); olfactory receptor family 52 subfamily N member 5; olfactory receptor OR11-33; Olfactory receptor OR11-62; olfactory receptor OR11-64; Olfactory receptor OR11-70; olfactory receptor OR11-8; olfactory receptor, family 52, subfamily A, member 5; olfactory receptor, family 52, subfamily B, member 2; olfactory receptor, family 52, subfamily K, member 1; olfactory receptor, family 52, subfamily N, member 4; olfactory receptor, family 52, subfamily N, member 5; OR11-33; OR11-62; OR11-64; OR11-70; OR11-8; OR52A5; OR52B2; OR52K1; OR52N4; OR52N5; OR52N5Q
Gene Symbols OR52Z1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR52Z1(Accession P0C646), corresponding to amino acid residues V260-C298.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283110
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.