Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR5A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549436
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549436 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549436 Supplier Invitrogen™ Supplier No. PA549436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR5A1 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR5A1
Gene Accession No. Q8NGJ0
Gene Alias Olfactory receptor 5A1; olfactory receptor family 5 subfamily A member 1; olfactory receptor OR11-249; olfactory receptor, family 5, subfamily A, member 1 pseudogene; OR11-249; OR5A1; OR5A1P; OST181
Gene Symbols OR5A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 301-335 amino acids from the C-terminal region of human OR5A1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.