Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR5AC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549254
Description
OR5AC2 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
OR5AC2 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9NZP5 | |
OR5AC2 | |
KLH conjugated synthetic peptide between 246-274 amino acids from the C-terminal region of human OR5AC2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR5AC2 | |
HSA1; Olfactory receptor 5AC2; olfactory receptor family 5 subfamily AC member 2; olfactory receptor, family 5, subfamily AC, member 2; OR5AC2 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
81050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction