Learn More
Description
Specifications
Specifications
| Antigen | OR5AU1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | olfactory receptor 5AU1, Olfactory receptor OR14-38, olfactory receptor, family 5, subfamily AU, member 1, OR14-38 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR5AU1 (NP_001004731). Peptide sequence LRPRSSYSLTQDRTVAVIYTVVIPVLNPLMYSLRNKDVKKALIKVWGRKT |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
