Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR5B12 Monoclonal Antibody (609CT8.2.4)
GREENER_CHOICE

Catalog No. PIMA524723
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524723 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524723 Supplier Invitrogen™ Supplier No. MA524723
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OR5B12 Monoclonal Antibody for Western Blot

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5B12
Applications Western Blot
Classification Monoclonal
Clone 609CT8.2.4
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR5B12
Gene Accession No. Q96R08
Gene Alias Olfactory receptor 5B12; olfactory receptor 5B16; olfactory receptor family 5 subfamily B member 12; olfactory receptor OR11-241; olfactory receptor, family 5, subfamily B, member 12 pseudogene; olfactory receptor, family 5, subfamily B, member 16; OR11-241; OR5B12; OR5B12P; OR5B16; OST743
Gene Symbols OR5B12
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 278-307 amino acids from the C-terminal region of human OR5B12.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.