Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR5D13 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306140APC
Description
OR5D13 Polyclonal antibody specifically detects OR5D13 in Human samples. It is validated for Western BlotSpecifications
OR5D13 | |
Polyclonal | |
Western Blot | |
olfactory receptor 5D13, Olfactory receptor OR11-142, Olfactory receptor OR11-148, olfactory receptor, family 5, subfamily D, member 13 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human OR5D13. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
390142 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction