Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR5D16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561292 Supplier Thermo Scientific Supplier No. PA561292
Add to Cart
Add to Cart

OR5D16 Polyclonal Antibody

OR5D16 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5D16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR5D16
Gene Accession No. Q8NGK9
Gene Alias Olfactory receptor 5D16; olfactory receptor family 5 subfamily D member 16; olfactory receptor OR11-154; olfactory receptor, family 5, subfamily D, member 16; OR11-154; OR5D16
Gene Symbols OR5D16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR5D16. Recombinant protein control fragment (Product #RP-100627).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.