Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR5H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548731
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548731 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548731 Supplier Invitrogen™ Supplier No. PA548731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR5H2 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5H2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR5H2
Gene Accession No. Q8NGV7
Gene Alias Olfactory receptor 5H2; olfactory receptor family 5 subfamily H member 2; olfactory receptor OR3-10; olfactory receptor, family 5, subfamily H, member 2; OR3-10; OR5H2
Gene Symbols OR5H2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 283-312 amino acids from the C-terminal region of human OR5H2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.