Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR5M10 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | OR5M10 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
OR5M10 Polyclonal specifically detects OR5M10 in Human samples. It is validated for Western Blot.Specifications
OR5M10 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
olfactory receptor 5M10, olfactory receptor, family 5, subfamily M, member 10, OR11-207 | |
The immunogen is a synthetic peptide directed towards the middle region of Human OR5M10 (NP_001004741). Peptide sequence NGLSQTLLTFHLSFCGSLEINHFYCADPPLIMLACSDTRVKKMAMFVVAG | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
390167 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title