Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR5R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571204 Supplier Invitrogen™ Supplier No. PA571204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5R1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR5R1
Gene Accession No. Q8NH85
Gene Alias Olfactory receptor 5R1; olfactory receptor family 5 subfamily R member 1 (gene/pseudogene); olfactory receptor OR11-185; olfactory receptor, family 5, subfamily R, member 1 pseudogene; OR11-185; OR5R1; OR5R1P
Gene Symbols OR5R1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human OR5R1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219479
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.