Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR5V1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571189
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 86%; Rabbit: 93%; Rat: 92%.
Odorant receptor.
Specifications
OR5V1 | |
Polyclonal | |
Unconjugated | |
OR5V1 | |
6M1-21; hs6M1-21; Olfactory receptor 5V1; olfactory receptor family 5 subfamily V member 1; olfactory receptor OR6-26; olfactory receptor, family 5, subfamily V, member 1; OR5V1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
81696 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UGF6 | |
OR5V1 | |
synthetic peptide directed towards the C-terminal region of human OR5V1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction