Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR6B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567744 Supplier Invitrogen™ Supplier No. PA567744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR6B1 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR6B1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR6B1
Gene Accession No. O95007
Gene Alias olfactory receptor 6B1; Olfactory receptor 7-3; olfactory receptor family 6 subfamily B member 1; olfactory receptor OR7-9; olfactory receptor, family 6, subfamily B, member 1; OR6B1; OR7-3; OR7-9
Gene Symbols OR6B1
Host Species Rabbit
Immunogen Synthesized peptide derived from human OR6B1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135946
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.