Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR6B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599924
Description
Antibody detects endogenous levels of total OR6B1.
Odorant receptor.
Specifications
OR6B1 | |
Polyclonal | |
Unconjugated | |
OR6B1 | |
olfactory receptor 6B1; Olfactory receptor 7-3; olfactory receptor family 6 subfamily B member 1; olfactory receptor OR7-9; olfactory receptor, family 6, subfamily B, member 1; OR6B1; OR7-3; OR7-9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
135946 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95007 | |
OR6B1 | |
A synthesized peptide derived from human OR6B1(Accession O95007), corresponding to amino acid residues F207-Y257. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction