Learn More
Invitrogen™ OR6K6 Polyclonal Antibody

Description
OR6K6 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | OR6K6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OR6K6 |
| Gene Accession No. | Q8NGW6 |
| Gene Alias | olfactory receptor 6K6; olfactory receptor family 6 subfamily K member 6; olfactory receptor OR1-21; olfactory receptor, family 6, subfamily K, member 6; OR1-21; OR6K6 |
| Gene Symbols | OR6K6 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.