Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR6X1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326089JF549
Description
OR6X1 Polyclonal antibody specifically detects OR6X1 in Human samples. It is validated for Western BlotSpecifications
OR6X1 | |
Polyclonal | |
Western Blot | |
olfactory receptor 6X1, olfactory receptor, family 6, subfamily X, member 1, OR11-270 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OR6X1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
390260 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction