Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR7D4 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP136975

Catalog No. NBP136975


Only null left
Add to Cart

Description

Description

OR7D4 Polyclonal specifically detects OR7D4 in Hamster (Negative) samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications

Specifications

OR7D4
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
hg105, Odorant receptor family subfamily D member 4RT, olfactory receptor 7D4, Olfactory receptor OR19-7, olfactory receptor, family 7, subfamily D, member 4, olfactory receptor, family 7, subfamily D, member 4 pseudogene, OR19-7, OR19B, OR19-Bodorant receptor family 7 subfamily D member 4 RT, OR7D4P
Goat
Affinity Purified
RUO
125958
Hamster (Negative)
IgG
Immunocytochemistry, Immunofluorescence, ELISA
0.5 mg/mL
Immunocytochemistry/Immunofluorescence 1:200, Peptide ELISA Detection limit 1:32000
NP_001005191.1
OR7D4
Peptide with sequence C-SIQARSKDISY corresponding to internal region according to NP_001005191.1.
0.1 mg
Primary
This antibody recognizes and epitope within the internal region. Hamster (Negative).
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only