Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR8B2/OR8B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103239 Supplier Invitrogen™ Supplier No. PA5103239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR8B2/B3.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR8B2/OR8B3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR8B3
Gene Accession No. Q8NGG8, Q96RD0
Gene Alias Olfactory receptor 8B2; olfactory receptor 8B3; olfactory receptor family 8 subfamily B member 2; olfactory receptor family 8 subfamily B member 3; olfactory receptor OR11-309; olfactory receptor OR11-310; olfactory receptor OR11-311; olfactory receptor, family 8, subfamily B, member 2; olfactory receptor, family 8, subfamily B, member 3; OR11-309; OR11-310; OR11-311; OR8B2; OR8B3
Gene Symbols OR8B2, OR8B3
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR8B2(Accession Q96RD0), corresponding to amino acid residues I207-F257.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26595, 390271
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.