Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR8D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2833440.1ML
Description
OR8D1 Polyclonal specifically detects OR8D1 in Human samples. It is validated for Western Blot.Specifications
OR8D1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
283159 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
JCG9, olfactory receptor 8D1, Olfactory receptor 8D3, Olfactory receptor OR11-301, olfactory receptor, family 8, subfamily D, member 1, olfactory receptor, family 8, subfamily D, member 3, Olfactory receptor-like protein JCG9, olfactory-like receptor JCG9, OR8D3, OST004PDJ9J14 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR8D1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction