Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR8D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571207 Supplier Thermo Scientific Supplier No. PA571207
Add to Cart
Add to Cart

OR8D2 Polyclonal Antibody

OR8D2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR8D2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR8D2
Gene Accession No. Q9GZM6
Gene Alias JCG2; olfactory receptor 8D2; olfactory receptor family 8 subfamily D member 2 (gene/pseudogene); olfactory receptor OR11-303; olfactory receptor, family 8, subfamily D, member 2; olfactory receptor-like protein JCG2; OR8D2
Gene Symbols OR8D2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human OR8D2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283160
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.