Learn More
Orai1 Antibody (6D11A11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP175523
Description
Orai1 Monoclonal specifically detects Orai1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Orai1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml | |
ORAI1 | |
Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1. | |
Affinity Purified | |
RUO | |
84876 | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
6D11A11 | |
Unconjugated | |
CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1 | |
Mouse | |
50 kDa | |
0.1 mg | |
Primary | |
ORAI1 antibody is predicted to have no cross-reactivity to ORAI2 or ORAI3. | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |