Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565926 Supplier Invitrogen™ Supplier No. PA565926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSAELNVPIDP SAPACPEPGH KGMDYRDWVR RSYLELVTSN HHSVQALSW Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI2 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. Like ORAI1, ORAI2 has been shown to function as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor, although at a lesser efficacy than ORAI1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORAI2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ORAI2
Gene Accession No. Q96SN7
Gene Alias A730041O15Rik; C7orf19; calcium release-activated calcium channel 2; calcium release-activated calcium modulator 2 protein; CAP-binding protein complex interacting protein 2; CAP-binding protein complex-interacting protein 2; CBCIP2; H_NH0514P08.8; MEM142B; ORAI calcium release-activated calcium modulator 2; ORAI2; PP1729; Protein orai-2; putative protein ORAI2-2; RGD1310213; TMEM142B; Transmembrane protein 142B
Gene Symbols ORAI2
Host Species Rabbit
Immunogen Recombinant Human ORAI2. Recombinant protein control fragment (Product #RP-106605).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.