Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORAI3 Antibody (1B4F1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | ORAI3 |
---|---|
Clone | 1B4F1 |
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
ORAI3 Monoclonal specifically detects ORAI3 in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
ORAI3 | |
1 mg/mL | |
Monoclonal | |
Mouse | |
Human, Rat | |
ORAI calcium release-activated calcium modulator 3, TMEM142C | |
ORAI3 | |
IgG2b | |
Affinity Purified |
1B4F1 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
93129 | |
A 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title