Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORAI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520370
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520370 Supplier Invitrogen™ Supplier No. PA520370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is A-20 cell lysate. PA5-2037°Can be used with blocking peptide PEP-0487.

ORAI3 is a protein coding gene that is involved in calcium signaling and regulation of calcium homeostasis. It is a member of the ORAI family of proteins that form calcium release-activated calcium (CRAC) channels in the plasma membrane of cells. These channels are activated by depletion of calcium from the endoplasmic reticulum, leading to an influx of calcium ions into the cytoplasm. ORAI3 has been implicated in a variety of physiological processes, including T-cell activation, muscle contraction, and neurotransmitter release. Dysregulation of ORAI3 expression or function has been associated with various diseases, including immune disorders, cancer, and cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORAI3
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ORAI3
Gene Accession No. Q6AXR8, Q6P8G8, Q9BRQ5
Gene Alias 9930124N15; BC061259; calcium release-activated calcium channel 3; calcium release-activated calcium modulator 3 protein; ORAI calcium release-activated calcium modulator 3; ORAI3; Protein orai-3; RGD1306538; Tmem142c; Transmembrane protein 142C
Gene Symbols ORAI3
Host Species Rabbit
Immunogen A 15 amino acid peptide from near the amino terminus of human ORAI3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269999, 309000, 93129
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.