Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sol-M™ Oral Syringe (Bulk, Non-Sterile)

Catalog No. 22024338
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
Color:
Amber
Clear
This item is not returnable. View return policy
This item is not returnable. View return policy

Oral syringe specifically for use with oral medications and featuring a gasket design.

  • Large variety of sizes and colors
  • Clear graduation markings for accurate medication delivery
  • Not made with natural rubber latex
  • Special tip design will not accept Luer connectors, preventing medication errors and the accidental intravenous injections of oral liquids
  • Optional amber barrel designed for light-sensitive drugs, in compliance with United States Pharmacopeia (USP) requirements

Specifications

Autoclavable Not Autoclavable
Material Polypropylene
Color Clear
Needle Gauge No Needle Included
Includes Tip Cap
Latex Free Yes
Disposable Single-use
Packaging Blister
Volume (Metric) 1 mL
For Use With (Equipment) Oral Syringe
Connection Type Catheter
Graduated Yes
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.