Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ORAOV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101219
Description
Antibody detects endogenous levels of total ORAOV1.
ORAOV1 is a protein coding gene. The complex LTO1:YAE1 functions as a target specific adapter that probably recruits apo-ABCE1 to the cytosolic iron-sulfur protein assembly (CIA) complex machinery. May be required for biogenesis of the large ribosomal subunit and initiation of translation. May play a role in the regulation of proline metabolism and ROS production.
Specifications
ORAOV1 | |
Polyclonal | |
Unconjugated | |
ORAOV1 | |
2210010N10Rik; Lto1; oral cancer overexpressed 1; oral cancer overexpressed protein 1-A; oral cancer-overexpressed protein 1; Oral cancer-overexpressed protein 1 homolog; Oraov1; Protein LTO1 homolog; TAOS1; Tumor-amplified and overexpressed sequence 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
220064, 309136 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WV07 | |
ORAOV1 | |
A synthesized peptide derived from human ORAOV1(Accession Q8WV07), corresponding to amino acid residues E83-S133. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction