Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORAOV1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549859

Catalog No. PA549859


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ORAOV1 protein.

ORAOV1 is a protein coding gene. The complex LTO1:YAE1 functions as a target specific adapter that probably recruits apo-ABCE1 to the cytosolic iron-sulfur protein assembly (CIA) complex machinery. May be required for biogenesis of the large ribosomal subunit and initiation of translation. May play a role in the regulation of proline metabolism and ROS production.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ORAOV1
Polyclonal
Unconjugated
ORAOV1
2210010N10Rik; Lto1; oral cancer overexpressed 1; oral cancer overexpressed protein 1-A; oral cancer-overexpressed protein 1; Oral cancer-overexpressed protein 1 homolog; Oraov1; Protein LTO1 homolog; TAOS1; Tumor-amplified and overexpressed sequence 1
Rabbit
Antigen affinity chromatography
RUO
220064
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8WV07
ORAOV1
Synthesized peptide derived from internal of human ORAV1 (aa 81-130).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.