Learn More
MilliporeSigma™ Supelco™ ORBO™ 1103 Porapak-Q (50/80), 150/75 mg W,W,W separators
Catalog No. 111009022 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
100 mm
Catalog No. Length (Metric)
11-100-9022 100 mm
1 options
Catalog No. 11-100-9022 Supplier MilliporeSigma™ Supelco™ Supplier No. 20063
Only null left
Add to Cart
Add to Cart
Used for gas and vapor sampling
ORBO™ 1103 Porapak™-Q (50/80), 150/75 mg solvent desorption tube is designed for gas and vapor sampling. It contains two beds of the same selective adsorbent separated by glass wool or foam. Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
TRUSTED_SUSTAINABILITY
Specifications
Material W,W,W Separators
Quantity 50/Pk.
Length (Metric) 100 mm
Diameter (Metric) 6 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC