Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ ORBO™ 1500 Precleaned Small PUF/Amberlite XAD-2/PUF Cartridge

Catalog No. 111004233 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
3/Pk.

Used for gas and vapor sampling

ORBO™-1500 cartridges are recommended for Pesticide/PCB applications. Sampling Rate: 1-5 L/min, PUF Density: 0.022 g/cm3
TRUSTED_SUSTAINABILITY

Specifications

Description PUF/XAD-2/PUF Layers (Each PUF 30 mm L; 1.5 g Amberlite XAD-2)
Quantity 3/Pk.
Length (Metric) 10 cm
Diameter (Metric) 22 mm

ORBO is a trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.