Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORC1 Antibody (7F6/1), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308793AF647
Description
ORC1 Monoclonal antibody specifically detects ORC1 in Human samples. It is validated for ELISASpecifications
ORC1 | |
Monoclonal | |
Alexa Fluor 647 | |
HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
Recombinant full-length human ORC1 protein (Uniprot: Q13415) | |
100 μg | |
Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4998 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
7F6/1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction