Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | ORC1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224600SS
![]() |
Novus Biologicals
NBP224600SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224600
![]() |
Novus Biologicals
NBP224600 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
ORC1 Polyclonal specifically detects ORC1 in Human, Mouse samples. It is validated for Western Blot.Specifications
ORC1 | |
Western Blot 1-3 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
ORC1 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, 89% homologous in dog and 84% homologous in mouse and rat. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
4998 | |
A portion of amino acids 170 to 220 of human ORC1L was used as immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title