Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORC1 Mouse anti-Human, Clone: 7F6/1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30747220UG
Description
ORC1 Monoclonal specifically detects ORC1 in Human samples. It is validated for ELISA.Specifications
ORC1 | |
Monoclonal | |
0.2 mg/ml | |
ELISA | |
HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
Recombinant full-length human ORC1 protein | |
20 μg | |
Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4998 | |
Store at 4°C. | |
IgG1 κ |
ELISA | |
7F6/1 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction