Learn More
Invitrogen™ ORC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590024
Description
Immunogen sequence: EDDAIQLVAR KVAALSGDAR RCLDICRRAT EICEFSQQKP DSPGLVTIAH SMEAVDEMFS SSYITAIKNS SVLEQSFLRA ILAEFRRSGL EEATFQQIYS QHVALCRMEG LPYPTMSETM AVCSHLGSCR LLLVEPSRND LLLRVRLNVS QDDVLYALKD E; Positive Samples: HeLa, 293T, A431, K562, Jurkat, MCF7, A549; Cellular Location: Nucleus.
The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is the largest subunit of the ORC complex. While other ORC subunits are stable throughout the cell cycle, the levels of this protein vary during the cell cycle, which has been shown to be controlled by ubiquitin-mediated proteolysis after initiation of DNA replication. This protein is found to be selectively phosphorylated during mitosis. It is also reported to interact with MYST histone acetyltransferase 2 (MyST2/HBO1), a protein involved in control of transcription silencing.
Specifications
ORC1 | |
Polyclonal | |
Unconjugated | |
Orc1 | |
AA545195; HS; HSORC1; MmORC1; orc1; orc1.L; Orc1l; origin recognition complex protein 1; origin recognition complex subunit 1; origin recognition complex subunit 1 L homeolog; origin recognition complex, subunit 1; origin recognition complex, subunit 1 homolog; origin recognition complex, subunit 1-like; PARC1; Replication control protein 1; XELAEV_18023178mg; xorc1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4998 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.40 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q13415 | |
Orc1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 696-856 of human ORC1 (NP_0011777481). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.