Learn More
Orc2 Antibody (C.15200198), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259237
Catalog No. NBP259237
Description
Description
Orc2 Monoclonal specifically detects Orc2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Orc2
Monoclonal
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
ORC2
The exact immunogen is propietary information.
50 μg
Cell Cycle and Replication
4999
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot
C.15200198
Western Blot 1:500
ORC2Lorigin recognition complex subunit 2, origin recognition complex protein 2 homolog, origin recognition complex, subunit 2, origin recognition complex, subunit 2 (yeast homolog)-like, origin recognition complex, subunit 2 homolog, origin recognition complex, subunit 2 homolog (yeast), origin recognition complex, subunit 2-like, origin recognition complex, subunit 2-like (yeast)
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only